Hexacarbonylmolybdenum ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Molybdenum carbonylMolybdenum hexacarbonylHexacarbonylmolybdenum(0)

42-1350-100G 42-1350-500G 42-1350-25G
100197-510EA 206.65 USD
100197-510 100208-432 100208-430
Hexacarbonylmolybdenum ≥98%
Hexacarbonylmolybdenum

Metal Carbonyls, Volatile Metal Carbonyls for CVD & ALD, Volatile Precursors for CVD

Formula: C₆MoO₆
MW: 264 g/mol
Melting Pt: 150…151 °C (dec.)
MDL Number: MFCD00003466
CAS Number: 13939-06-5
UN: 3466
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR