Cyclopentadienylmolybdenum tricarbonyl dimer ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Bis(tricarbonylcyclopentadienylmolybdenum)
42-0400-25G
42-0400-5G
100205-122EA
175.51
USD
100205-122
100202-402
Cyclopentadienylmolybdenum tricarbonyl dimer ≥98%
Cyclopentadienylmolybdenum tricarbonyl dimer
Metallocenes, Derivatives & Cp Precursors
Formula:
C₁₆H₁₀Mo₂O₆ MW: 490.13 g/mol Melting Pt: 220 °C (dec.) |
MDL Number:
MFCD00009947 CAS Number: 12091-64-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...