Trimethylarsine ≥99%

Supplier: STREM CHEMICALS INC MS

Danger

33-3750.5 33-3750.25
100209-480EA 539.26 USD InStock
100209-480 100202-662
Trimethylarsine ≥99%
Trimethylarsine

Metal Alkyls, Volatile Precursors for CVD

Formula: C₃H₉As
MW: 120,025 g/mol
Boiling Pt: 51 °C
Melting Pt: –87,3 °C
Density: 1,124
Flash Pt: 100 °F
Storage Temperature: ambient
MDL Number: MFCD00014838
CAS Number: 593-88-4
EINECS: 209-815-8
UN: 1992
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR