Triethylarsine ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

33-3400-5G 33-3400-25G
100205-930EA 1755.08 USD
100205-930 100207-386
Triethylarsine ≥99%
Triethylarsine

Metal Alkyls, Volatile Precursors for CVD

Formula: C₆H₁₅As
MW: 162.105 g/mol
Boiling Pt: 140 °C
Melting Pt: –91 °C
Density: 1.152
MDL Number: MFCD00015093
CAS Number: 617-75-4
UN: 1992
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR