Dimethylzinc 10% (w/w) in hexanes ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Sure/Seal™ bottle (Trademark of Sigma-Aldrich Co. LLC)
30-5062-250G
30-5062-50G
100203-094EA
427.45
USD
100203-094
100203-092
Dimethylzinc 10% (w/w) in hexanes ≥99%
Dimethylzinc
Formula:
C₂H₆Zn |
MDL Number:
MFCD00014854 CAS Number: 544-97-8 UN: 1993 ADR: 3,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...