Zinc trifluoromethanesulfonate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Zinc(II) trifluoromethanesulfonate
, Zinc trifluoromethanesulphonate
, Zinc(II) trifluoromethanesulphonate
, Zinc trifluoromethanesulfonate
, Zinc triflate
30-4000-5G
30-4000-25G
100204-040EA
46.71
USD
100204-040
100203-766
Zinc trifluoromethanesulfonate ≥98%
Zinc trifluoromethanesulphonate
Metal Triflate
Formula:
C₂F₆O₆S₂Zn MW: 363.53 g/mol |
MDL Number:
MFCD00013229 CAS Number: 54010-75-2 UN: 3260 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...