Dicarbonylbis(triphenylphosphine)nickel ≥98%
Supplier: STREM CHEMICALS INC MS
Synonyms:
Bis(triphenylphosphine)dicarbonylnickel
, Bis(triphenylphosphine)nickel dicarbonyl
28-0100-5G
28-0100-25G
28-0100-100G
100202-492EA
693.54
USD
100202-492
100208-410
100208-408
Dicarbonylbis(triphenylphosphine)nickel ≥98%
Dicarbonylbis(triphenylphosphine)nickel
Formula:
C₃₈H₃₀NiO₂P₂ MW: 639.29 g/mol Melting Pt: 210…215 °C (dec.) |
MDL Number:
MFCD00009591 CAS Number: 13007-90-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...