1,3-Bis(diphenylphosphino)propanenickel(II) chloride ≥99%
Supplier: STREM CHEMICALS INC MS
Synonyms:
Dichloro[bis(1,3-diphenylphosphino)propane]nickel(II)
, Dichloro[1,3-bis(diphenylphosphino)propane]nickel(II)
, 1,3-Bis(diphenylphosphino)propane nickel(II)chloride
, [1,3-Bis(diphenylphosphino)propane]nickel (II) chloride
, [1,3-Bis(diphenylphosphino)propane]nickel(II) dichloride
, [1,3-Bis(diphenylphosphino)propane]dichloronickel(II)
28-0080-25G
28-0080-5G
100197-476EA
104.74
USD
100197-476
100197-474
1,3-Bis(diphenylphosphino)propanenickel(II) chloride ≥99%
1,3-Bis(diphenylphosphino)propanenickel(II) chloride
Formula:
C₂₇H₂₆Cl₂NiP₂ MW: 542.042 g/mol |
MDL Number:
MFCD00015318 CAS Number: 15629-92-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...