Cobalt(II) Phthalocyanine
Supplier: STREM CHEMICALS INC MS
Synonyms:
Phthalocyanine Cobalt(II)
, 29H,31H-Phthalocyanine, cobalt complex
, Cobalt(II) phthalocyanine
27-0488-1G
27-0488-5G
100208-010EA
199.57
USD
100208-010
100205-060
Cobalt(II) Phthalocyanine
Cobalt(II) Phthalocyanine
Formula:
C₃₂H₁₆CoN₈ MW: 571.47 g/mol |
MDL Number:
MFCD00010718 CAS Number: 3317-67-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...