[5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrinato]cobalt(II) ≥96%
Supplier: STREM CHEMICALS INC MS
Synonyms:
Cobalt tetramethoxyphenylporphyrin
, Cobalt(II) meso-tetra(4-methoxyphenyl)porphine
, [5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphinato]cobalt(II)
, 5,10,15,20-Tetrakis (4-methoxyphenyl)-21H,23H-porphine cobalt (II)
27-0465-1G
27-0465-5G
100199-842EA
304.31
USD
100199-842
100199-840
[5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrinato]cobalt(II) ≥96%
[5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrinato]cobalt(II)
Porphine
Formula:
C₄₈H₃₆CoN₄O₄ MW: 791.77 g/mol |
MDL Number:
MFCD00010724 CAS Number: 28903-71-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...