(S)-(+)-1-[(R)-2-(Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine ethanol adduct ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
(2S)-1-[(1S)-1-(Dicyclohexylphosphino)ethyl]-2-(diphenylphosphino)ferrocene
, (S)-(R)-JOSIPHOS
26-1211-100MG
26-1211-500MG
100204-588EA
530.77
USD
100204-588
100209-840
(S)-(+)-1-[(R)-2-(Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine ethanol adduct ≥97%
(S)-(+)-1-[(R)-2-(Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine ethanol adduct
JOSIPHOS, Phosphine
Formula:
C₃₆H₄₄FeP₂ MW: 594.5300 g/mol |
MDL Number:
MFCD00800285 CAS Number: 162291-02-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...