(R)-(-)-1-[(S)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldicyclohexylphosphine ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
(R)-1-[(R)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldicyclohexylphosphine
26-1000-100MG
26-1000-500MG
100204-566EA
530.77
USD
100204-566
100209-818
(R)-(-)-1-[(S)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldicyclohexylphosphine ≥97%
(R)-(-)-1-[(S)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldicyclohexylphosphine
JOSIPHOS, Phosphine
Formula:
C₃₆H₅₆FeP₂ MW: 606.6300 g/mol |
MDL Number:
MFCD01074441 CAS Number: 167416-28-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...