(Indenyl)titanium(IV) trichloride ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Indenyltitanium trichloride
, Trichloro(indenyl)titanium(IV)
22-0950-25G
22-0950-1G
22-0950-5G
100201-530EA
141.54
USD
100201-530
100197-444
100203-042
(Indenyl)titanium(IV) trichloride ≥99%
(Indenyl)titanium(IV) trichloride
Metallocenes, Derivatives & Cp Precursors
Formula:
C₉H₇Cl₃Ti MW: 269.37 g/mol |
MDL Number:
MFCD00269849 CAS Number: 84365-55-9 UN: 1759 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...