(4R,5R)-(-)-2,2-Dimethyl-α,α,α',α'-tetra(1-naphthyl)-1,3-dioxolane-4,5-dimethanolatotitanium(IV) dichloride acetonitrile adduct
Supplier: STREM CHEMICALS INC MS
|
Warning
|
22-0761-1G
22-0761-250MG
100203-036EA
665.23
USD
100203-036
100203-034
(4R,5R)-(-)-2,2-Dimethyl-α,α,α',α'-tetra(1-naphthyl)-1,3-dioxolane-4,5-dimethanolatotitanium(IV) dichloride acetonitrile adduct
(4R,5R)-(-)-2,2-Dimethyl-α,α,α',α'-tetra(1-naphthyl)-1,3-dioxolane-4,5-dimethanolatotitanium(IV) dichloride acetonitrile adduct
Color and Form: yellow pwdr.
Stability: air sensitive, moisture sensitive
Formula:
C₄₇H₃₆Cl₂O₄Ti |
MDL Number:
MFCD05861615 CAS Number: 197389-47-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...