Cyclopentadienyltitanium trichloride ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
22-0400-1G
22-0400-5G
22-0400-25G
100205-010EA
65.11
USD
100205-010
100208-378
100208-376
Cyclopentadienyltitanium trichloride ≥99%
Cyclopentadienyltitanium trichloride
Metallocenes, Derivatives & Cp Precursors
Formula:
C₅Cl₃Ti MW: 214,28 g/mol Melting Pt: 198…200 °C |
UN:
1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...