Bis(cyclopentadienyl)titanium dichloride ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Titanocene dichloride
, Bis(cyclopentadienyl)titanium(IV) dichloride
, Titanocenedichloride
22-0200-25G
22-0200-100G
100203-032EA
339.69
USD
100203-032
100198-152
Bis(cyclopentadienyl)titanium dichloride ≥99%
Bis(cyclopentadienyl)titanium dichloride
Metallocenes, Derivatives & Cp Precursors
Formula:
C₁₀H₁₀Cl₂Ti MW: 248.97 g/mol Melting Pt: 289…291 °C |
MDL Number:
MFCD00003723 CAS Number: 1271-19-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...