Scandium (III) trifluoromethanesulfonate [∼13% Sc(SO₃CF₃)₃] on polystyrene, microencapsulated

Warning

Synonyms: Scandium(III) trifluoromethanesulphonateScandium triflateScandium(III) trifluoromethanesulfonate

21-2004-500MG
100203-754EA 622.71 USD
100203-754
Scandium (III) trifluoromethanesulfonate [∼13% Sc(SO₃CF₃)₃] on polystyrene, microencapsulated
Scandium (III) trifluoromethanesulphonate

Metal Triflate

Formula: C₃F₉O₉S₃Sc
MW: 492.17 g/mol
MDL Number: MFCD00192433
CAS Number: 144026-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR