Potassium hexafluoronickelate(IV) ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
19-2000-10G
19-2000-50G
100207-364EA
147.2
USD
100207-364
100196-920
Potassium hexafluoronickelate(IV) ≥99%
Potassium hexafluoronickelate (IV)
Formula:
F₆K₂Ni MW: 250.88 g/mol |
MDL Number:
MFCD00083064 CAS Number: 17218-47-2 UN: 3288 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...