(S)-(+)-5,5',6,6',7,7',8,8'-Octahydro-3,3'-di-t-butyl-1,1'-bi-2-naphthol dipotassium salt
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Potassium (S)-3,3'-di-tert-butyl-5,5',6,6',7,7',8,8'-octahydro-[1,1'-binaphthalene]-2,2'-bis(olate)
19-1601-100MG
19-1601-500MG
100209-756EA
212.31
USD
100209-756
100209-754
(S)-(+)-5,5',6,6',7,7',8,8'-Octahydro-3,3'-di-t-butyl-1,1'-bi-2-naphthol dipotassium salt
(S)-(+)-5,5',6,6',7,7',8,8'-Octahydro-3,3'-di-t-butyl-1,1'-bi-2-naphthol dipotassium salt
Formula:
C₂₈H₃₆O₂.2K MW: 482.78 g/mol |
MDL Number:
MFCD03788939 CAS Number: 821793-28-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...