Trimethylphosphonium tetrafluoroborate ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
15-6560-5G
15-6560-1G
100204-004EA
440.18
USD
100204-004
100204-002
Trimethylphosphonium tetrafluoroborate ≥99%
Trimethylphosphonium tetrafluoroborate
Formula:
C₃H₁₀BF₄P |
MDL Number:
MFCD03840581 CAS Number: 154358-50-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...