Trihexyl(tetradecyl)phosphonium decanoate ≥95% (CYPHOS® IL 103)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
15-6386-50G
15-6386-10G
100203-994EA
50.95
USD
100203-994
100203-996
Trihexyl(tetradecyl)phosphonium decanoate ≥95% (CYPHOS® IL 103)
Trihexyl(tetradecyl)phosphonium decanoate
CYPHOS IL, Ionic Liquids, Phosphine
Formula:
C₄₂H₈₇O₂P Melting Pt: 24 °C Density: 0,883 |
MDL Number:
MFCD04117698 CAS Number: 465527-65-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...