Triethylphosphonium tetrafluoroborate ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
15-6355-1G
15-6355-5G
100209-736EA
52.37
USD
100209-736
100209-738
Triethylphosphonium tetrafluoroborate ≥99%
Triethylphosphonium tetrafluoroborate
Formula:
C₆H₁₆BF₄P |
MDL Number:
MFCD03788938 CAS Number: 817575-08-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...