(S)-(-)-2,2',6,6'-Tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridine ≥95%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
(S)-4,4'-Bis(diphenylphosphino)-2,2',6,6'-tetramethoxy-3,3'-bipyridine
, CTH-(S)-P-Phos
15-5201-500MG
15-5201-100MG
100204-964EA
447.26
USD
100204-964
100204-962
(S)-(-)-2,2',6,6'-Tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridine ≥95%
(S)-(-)-2,2',6,6'-Tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridine
Phosphine, P-PHOS
Formula:
C₃₈H₃₄N₂O₄P₂ MW: 644.64 g/mol |
MDL Number:
MFCD04038734 CAS Number: 362524-23-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...