(R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridine ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
(R)-2,2',6,6'-Tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridine
, CTH-(R)-P-Phos
15-5200-100MG
15-5200-500MG
100204-960EA
447.26
USD
100204-960
100204-958
(R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridine ≥97%
(R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridine
Phosphine, P-PHOS
Formula:
C₃₈H₃₄N₂O₄P₂ MW: 644.64 g/mol |
MDL Number:
MFCD04038734 CAS Number: 221012-82-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...