(S)-(-)-7,7'-Bis(diphenylphosphino)-2,2',3,3'-tetrahydro-1,1'-spirobiindane ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (S)-SDP

15-5175-100MG
101418-704EA 370.83 USD
101418-704
(S)-(-)-7,7'-Bis(diphenylphosphino)-2,2',3,3'-tetrahydro-1,1'-spirobiindane ≥97%
(S)-(-)-7,7'-Bis(diphenylphosphino)-2,2',3,3'-tetrahydro-1,1'-spirobiindane

Phosphine, SDP

Melting Pt: 206…208 °C
MDL Number: MFCD08459344
CAS Number: 528521-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR