(R)-(+)-7,7'-Bis(diphenylphosphino)-2,2',3,3'-tetrahydro-1,1'-spirobiindane ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (R)-SDP

15-5174-100MG
101418-700EA 370.83 USD
101418-700
(R)-(+)-7,7'-Bis(diphenylphosphino)-2,2',3,3'-tetrahydro-1,1'-spirobiindane ≥97%
(R)-(+)-7,7'-Bis(diphenylphosphino)-2,2',3,3'-tetrahydro-1,1'-spirobiindane

Phosphine, SDP

Melting Pt: 200…202 °C
MDL Number: MFCD08459344
CAS Number: 917377-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR