(S)-(-)-7,7'-Bis[di(3,5-dimethylphenyl)phosphino]-2,2',3,3'-tetrahydro-1,1'-spirobiindane ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (S)-Xyl-SDP(S)-7,7'-Bis(bis(3,5-dimethylphenyl)phosphaneyl)-2,2',3,3'-tetrahydro-1,1'-spirobi[indene]

15-5169-100MG
101418-696EA 343.94 USD
101418-696
(S)-(-)-7,7'-Bis[di(3,5-dimethylphenyl)phosphino]-2,2',3,3'-tetrahydro-1,1'-spirobiindane ≥97%
(S)-(-)-7,7'-Bis[di(3,5-dimethylphenyl)phosphino]-2,2',3,3'-tetrahydro-1,1'-spirobiindane

Phosphine, SDP

Formula: C₄₉H₅₀P₂
MW: 700.8688 g/mol
MDL Number: MFCD08459343
CAS Number: 528521-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR