Dichloro(methyl)phosphine ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Methyldichlorophosphine

15-3220-5G
100208-326EA 363.75 USD
100208-326
Dichloro(methyl)phosphine ≥97%
Dichloro(methyl)phosphine

Phosphine

Formula: CH₃Cl₂P
MW: 116.91 g/mol
Boiling Pt: 81 °C
Density: 1.31
MDL Number: MFCD00013616
CAS Number: 676-83-5
UN: 2845
ADR: 4.2,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR