(S)-(-)-2,2'-Bis(diphenylphosphino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
(S)-(-)-H8-BINAP
15-2973-50MG
15-2973-250MG
101418-656EA
97.66
USD
101418-656
101418-658
(S)-(-)-2,2'-Bis(diphenylphosphino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl
(S)-(-)-2,2'-Bis(diphenylphosphino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl
BINAP, Phosphine
Formula:
C₄₄H₄₀P₂ MW: 630.7400 g/mol Melting Pt: 207…208 °C |
MDL Number:
MFCD01630795 CAS Number: 139139-93-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...