(R)-(+)-2,2'-Bis(diphenylphosphino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (R)-2,2'-Bis(diphenylphosphino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl(R)-(+)-H8-BINAP

15-2972-50MG 15-2972-250MG
101418-652EA 97.66 USD
101418-652 101418-654
(R)-(+)-2,2'-Bis(diphenylphosphino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl
(R)-(+)-2,2'-Bis(diphenylphosphino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl

BINAP, Phosphine

Formula: C₄₄H₄₀P₂
MW: 630.7400 g/mol
Melting Pt: 207…208 °C
MDL Number: MFCD01630795
CAS Number: 139139-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR