(R,S)-N-PINAP ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

15-1786-250MG
101418-644EA 171.26 USD
101418-644
(R,S)-N-PINAP ≥97%
(R,S)-N-PINAP

Phosphine

Formula: C₃₈H₃₀N₃P
MW: 559,64 g/mol
Melting Pt: >210 °C
MDL Number: MFCD09264274
CAS Number: 828927-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR