1-Diphenylphosphino-2-(N,N-dimethylamino)-1H-indene ≥99% (contains vinylic isomer)

Supplier: STREM CHEMICALS INC MS
Warning

15-1748-1G
101418-628EA 387.82 USD
101418-628
1-Diphenylphosphino-2-(N,N-dimethylamino)-1H-indene ≥99% (contains vinylic isomer)
1-Diphenylphosphino-2-(N,N-dimethylamino)-1H-indene

Phosphine

MDL Number: MFCD09264273
CAS Number: 628323-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR