(3aR,8aR)-(-)-(2,2-Dimethyl-4,4,8,8-tetraphenyl-tetrahydro-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepin-6-yl)dimethylamine ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
(3aR,8aR)-N,N,2,2-Tetramethyl-4,4,8,8-tetraphenyltetrahydro-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepin-6-amine
15-1505-500MG
15-1505-100MG
100197-074EA
179.75
USD
100197-074
100197-076
(3aR,8aR)-(-)-(2,2-Dimethyl-4,4,8,8-tetraphenyl-tetrahydro-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepin-6-yl)dimethylamine ≥98%
(3aR,8aR)-(-)-(2,2-Dimethyl-4,4,8,8-tetraphenyl-tetrahydro-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepin-6-yl)dimethylamine
MONOPHOS, Phosphine
Formula:
C₃₃H₃₄NO₄P MW: 539.6000 g/mol Melting Pt: 218…221 °C |
MDL Number:
MFCD04117685 CAS Number: 213843-90-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...