Chloro(dimethyl)phosphine ≥97%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Dimethylchlorophosphine
15-1250-1G
15-1250-5G
100199-790EA
260.43
USD
100199-790
100199-792
Chloro(dimethyl)phosphine ≥97%
Chloro(dimethyl)phosphine
Phosphine
Formula:
C₂H₆ClP MW: 96.5 g/mol Boiling Pt: 76…77 °C Density: 1.22 Flash Pt: –1 °F |
MDL Number:
MFCD00013620 CAS Number: 811-62-1 UN: 2845 ADR: 4.2,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...