2'-Dicyclohexylphosphino-2,6-dimethoxy-3-sulfonato-1,1'-biphenyl hydrate sodium salt ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Sodium 2'-(dicyclohexylphosphino)-2,6-dimethoxy-[1,1'-biphenyl]-3-sulfonate hydrate
, Sodium 2'-(dicyclohexylphosphino)-2,6-dimethoxy-[1,1'-biphenyl]-3-sulphonate hydrate
, (water soluble SPhos)
15-1142-500MG
15-1142-2G
101324-598EA
417.54
USD
101324-598
101324-596
2'-Dicyclohexylphosphino-2,6-dimethoxy-3-sulfonato-1,1'-biphenyl hydrate sodium salt ≥98%
2'-Dicyclohexylphosphino-2,6-dimethoxy-3-sulfonato-1,1'-biphenyl hydrate sodium salt
Buchwald Precatalysts & Ligands, Phosphine, Phosphine, Water Soluble
Formula:
C₂₆H₃₆NaO₆PS MW: 530.5900 g/mol |
MDL Number:
MFCD07781994 CAS Number: 1049726-96-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...