Dicyclohexyl[9-(3-phenylpropyl)-9-fluorenyl]phosphonium tetrafluoroborate ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Dicyclohexyl[9-(3-phenylpropyl)fluoren-9-yl]phosphonium tetrafluoroboratecataCXium® FPrPh

15-1076-2G
101433-132EA 445.85 USD
101433-132
Dicyclohexyl[9-(3-phenylpropyl)-9-fluorenyl]phosphonium tetrafluoroborate ≥95%
Dicyclohexyl[9-(3-phenylpropyl)-9-fluorenyl]phosphonium tetrafluoroborate

cataCXium

Formula: C₃₄H₄₂BF₄P
MW: 568.476 g/mol
MDL Number: MFCD10567004
CAS Number: 1007311-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR