(9-Benzyl-9-fluorenyl)dicyclohexylphosphonium tetrafluoroborate ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Dicyclohexyl(9-benzylfluoren-9-yl)phosphonium tetrafluoroborate
, cataCXium® FBn
15-1072-500MG
15-1072-2G
101433-124EA
445.85
USD
101433-124
101433-122
(9-Benzyl-9-fluorenyl)dicyclohexylphosphonium tetrafluoroborate ≥97%
(9-Benzyl-9-fluorenyl)dicyclohexylphosphonium tetrafluoroborate
cataCXium
Formula:
C₃₂H₃₈BF₄P MW: 540.422 g/mol |
MDL Number:
MFCD10567010 CAS Number: 937378-18-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...