2-Di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: tBuXPhos2-Di-tert-butylphosphino-2',4',6'-triisopropylbiphenylt-BuXPhosDi-tert-butyl(2',4',6'-triisopropyl-[1,1'-biphenyl]-2-yl)phosphine

15-1052-500MG 15-1052-2G
100206-346EA 39.63 USD
100206-346 100198-002
2-Di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl ≥98%
2-Di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl

Buchwald Precatalysts & Ligands, Phosphine

Formula: C₂₉H₄₅P
MW: 424.65 g/mol
Melting Pt: 147…149 °C
MDL Number: MFCD06411306
CAS Number: 564483-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR