Dicyclohexylchlorophosphine ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Chlorodicyclohexylphosphine

15-1050-1G 15-1050-5G
100200-870EA 380.74 USD
100200-870 100207-572
Dicyclohexylchlorophosphine ≥98%
Dicyclohexylchlorophosphine

Phosphine

Formula: C₁₂H₂₂ClP
Boiling Pt: 96…99 °C (0.1 mmHg)
Density: 1.054
MDL Number: MFCD00014294
CAS Number: 16523-54-9
UN: 1760
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR