Chloro(di-tert-butyl)phosphine ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Di-t-butylchlorophosphine
, Di-tert-butylchlorophosphine
15-1000-5G
15-1000-1G
100197-400EA
168.43
USD
100197-400
100202-972
Chloro(di-tert-butyl)phosphine ≥98%
Chloro(di-tert-butyl)phosphine
Phosphine
Formula:
C₈H₁₈ClP MW: 180.66 g/mol Boiling Pt: 69…70 °C (10 mmHg) Melting Pt: 2…3 °C Density: 0.951 Flash Pt: 142 °F |
MDL Number:
MFCD00008815 CAS Number: 13716-10-4 UN: 1760 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...