Di-1-adamantylphosphine ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

15-0954-250MG 15-0954-1G
100206-334EA 100.49 USD
100206-334 100206-336
Di-1-adamantylphosphine ≥97%
Di-1-adamantylphosphine

Phosphine

MDL Number: MFCD06658117
CAS Number: 131211-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR