(R,R)-1,2-Bis[(2-methoxyphenyl)phenylphosphino]ethane ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
(R,R)-Ethylenebis[(2-methoxyphenyl)phenylphosphine]
, (R,R)-DIPAMP
, (R,R)-(-)-1,2-Bis[(2-methoxyphenyl)(phenyl)phosphino]ethane
, (1R,2R)-Bis[(2-methoxyphenyl)phenylphosphino]ethane
, (-)-DIPAMP
15-0481-1G
15-0481-250MG
101324-586EA
1047.38
USD
101324-586
101324-584
(R,R)-1,2-Bis[(2-methoxyphenyl)phenylphosphino]ethane ≥98%
(R,R)-1,2-Bis[(2-methoxyphenyl)phenylphosphino]ethane
Phosphine
Formula:
C₂₈H₂₈O₂P₂ MW: 458.48 g/mol Melting Pt: 102…103 °C |
MDL Number:
MFCD05863546 CAS Number: 55739-58-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...