(R)-(+)-XylBINAP ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (R)-2,2'-Bis(di-3,5-xylylphosphino)-1,1'-binaphthyl(R)-(+)-2,2'-Bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl

15-0476-250MG
101418-572EA 67.94 USD
101418-572
(R)-(+)-XylBINAP ≥98%
(R)-(+)-XylBINAP

BINAP, Phosphine

Formula: C₅₂H₄₈P₂
MW: 734.90 g/mol
Melting Pt: 203…206 °C
MDL Number: MFCD01630821
CAS Number: 137219-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR