1,8-Bis(diphenylphosphino)octane ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
15-0420-5G
15-0420-1G
101418-562EA
243.45
USD
101418-562
101418-564
1,8-Bis(diphenylphosphino)octane ≥99%
1,8-Bis(diphenylphosphino)octane
Phosphine
Formula:
C₃₂H₃₆P₂ MW: 482.5800 g/mol |
MDL Number:
MFCD09834841 CAS Number: 41625-30-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...