Bis[2-(diphenylphosphino)phenyl]ether ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Bis(2-diphenylphosphinophenyl)ether
, (Oxybis(2,1-phenylene))bis(diphenylphosphine)
, 2,2'-Bis(diphenylphosphino)diphenyl Ether
, DPEphos
, (Oxydi-2,1-phenylene)bis(diphenylphosphine)
15-0380-5G
15-0380-25G
100206-320EA
93.42
USD
100206-320
100206-318
Bis[2-(diphenylphosphino)phenyl]ether ≥98%
Bis[2-(diphenylphosphino)phenyl]ether
Phosphine, Pincer Ligands and Complexes
Formula:
C₃₆H₂₈OP₂ MW: 538.57 g/mol Melting Pt: 181…184 °C |
MDL Number:
MFCD00233863 CAS Number: 166330-10-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...