(R)-5,5'-Bis(diphenylphosphino)-4,4'-bibenzo[d][1,3]dioxole ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
(R)-(+)-5,5'-Bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole
, (R)-(+)-SEGPHOS®
, (R)-(+)-4,4'-Bis(diphenylphosphino)-3,3'-bi(1,2-methylenedioxybenzene)
15-0136-250MG
101418-532EA
67.94
USD
101418-532
(R)-5,5'-Bis(diphenylphosphino)-4,4'-bibenzo[d][1,3]dioxole ≥98%
(R)-5,5'-Bis(diphenylphosphino)-4,4'-bibenzo[d][1,3]dioxole
Phosphine, SEGPHOS
Formula:
C₃₈H₂₈O₄P₂ MW: 610.570 g/mol Melting Pt: 168…172 °C |
MDL Number:
MFCD09753005 CAS Number: 244261-66-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...