(R)-(+)-2,2'-Bis(di-2-furanylphosphino)-6,6'-dimethoxy-1,1'-biphenyl ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
(R)-(6,6'-Dimethoxybiphenyl-2,2'-diyl)bis(di-2-furylphosphine)
15-0112-500MG
15-0112-100MG
101418-514EA
135.88
USD
101418-514
101418-516
(R)-(+)-2,2'-Bis(di-2-furanylphosphino)-6,6'-dimethoxy-1,1'-biphenyl ≥97%
(R)-(+)-2,2'-Bis(di-2-furanylphosphino)-6,6'-dimethoxy-1,1'-biphenyl
BIPHEP, Phosphine
Formula:
C₃₀H₂₄O₆P₂ MW: 542,45 g/mol |
MDL Number:
MFCD09753004 CAS Number: 145214-57-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...