1,2-Bis(di(pentafluorophenyl)phosphino)ethane ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
15-0094-5G
15-0094-1G
100202-940EA
148.62
USD
100202-940
100198-124
1,2-Bis(di(pentafluorophenyl)phosphino)ethane ≥99%
1,2-Bis(di(pentafluorophenyl)phosphino)ethane
Phosphine
Formula:
C₂₆H₄F₂₀P₂ Melting Pt: 188…190 °C |
MDL Number:
MFCD00058841 CAS Number: 76858-94-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...