Bis(dimethylphosphino)methane ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
15-0093-5G
15-0093-1G
100200-242EA
989.35
USD
100200-242
100200-240
Bis(dimethylphosphino)methane ≥98%
Bis(dimethylphosphino)methane
Phosphine
Formula:
C₅H₁₄P₂ Boiling Pt: 42 °C (12 mmHg) Density: 0.86 |
MDL Number:
MFCD00058731 CAS Number: 64065-08-3 UN: 2845 ADR: 4.2,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...