Bis(dichlorophosphino)methane ≥90%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Methylenebis(dichlorophosphine)
, dichloro(dichlorophosphanylmethyl)phosphane
, Phosphonous dichloride,P,P'-methylenebis-
15-0081-10G
15-0081-2G
100201-442EA
155.69
USD
100201-442
100201-440
Bis(dichlorophosphino)methane ≥90%
Bis(dichlorophosphino)methane
Phosphine
Formula:
CH₂Cl₄P₂ MW: 217.775 g/mol Boiling Pt: 101…105 °C (22 mmHg) Density: 1,601 |
MDL Number:
MFCD01073767 CAS Number: 28240-68-8 UN: 1760 ADR: 8,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...